Assay Details
| Gene Symbol: NIP5;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117106.3 | NP_192776.1 | 1 - 2 | 1126 | 74 | |
| GenBank mRNA | AY087560.1 | - | 1 - 2 | 552 | 74 | |
| BT030327.1 | - | 1 - 2 | 240 | 74 | ||
More Information
Set Membership: |
|

NIP5;1
Genomic Map