Assay Details
Gene Symbol: MTSSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117176.3 | NP_192844.2 | 4 - 5 | 659 | 104 | |
GenBank mRNA | AJ582222.1 | - | 3 - 4 | 431 | 104 | |
BT004166.1 | - | 4 - 5 | 596 | 104 | ||
BT005015.1 | - | 3 - 4 | 431 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
