Assay Details
| Gene Symbol: SGT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117197.4 | NP_192865.1 | 1 - 2 | 349 | 66 | |
| GenBank mRNA | AF370229.1 | - | 1 - 2 | 292 | 66 | |
| AF428340.1 | - | 1 - 2 | 287 | 66 | ||
| AF439976.1 | - | 1 - 2 | 156 | 66 | ||
| AJ318019.1 | - | 1 - 2 | 195 | 66 | ||
| AY085464.1 | - | 1 - 2 | 212 | 66 | ||
| AY150487.1 | - | 1 - 2 | 156 | 66 | ||
More Information
Set Membership: |
|

SGT1B
Genomic Map