Assay Details
| Gene Symbol: SGT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117197.4 | NP_192865.1 | 5 - 6 | 774 | 101 | |
| GenBank mRNA | AF370229.1 | - | 5 - 6 | 717 | 101 | |
| AF428340.1 | - | 5 - 6 | 712 | 101 | ||
| AF439976.1 | - | 5 - 6 | 581 | 101 | ||
| AJ318019.1 | - | 5 - 6 | 620 | 101 | ||
| AY085464.1 | - | 5 - 6 | 637 | 101 | ||
| AY150487.1 | - | 5 - 6 | 581 | 101 | ||
More Information
Set Membership: |
|

SGT1B
Genomic Map