Assay Details
| Gene Symbol: AT4G11290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117200.4 | NP_192868.1 | 3 - 4 | 704 | 99 | |
| GenBank mRNA | AK221643.1 | - | 3 - 4 | 614 | 99 | |
| BT026448.1 | - | 3 - 4 | 574 | 99 | ||
More Information
Set Membership: |
|

AT4G11290
Genomic Map