Assay Details
| Gene Symbol: RHA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117207.3 | NP_192875.1 | 1 | 684 | 104 | |
| GenBank mRNA | AY065243.1 | - | 1 | 513 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RHA1B| Gene Symbol: RHA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117207.3 | NP_192875.1 | 1 | 684 | 104 | |
| GenBank mRNA | AY065243.1 | - | 1 | 513 | 104 | |
Set Membership: |
|