Assay Details
| Gene Symbol: AT-HSFB2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117235.5 | NP_192903.1 | 1 - 2 | 1000 | 117 | |
| GenBank mRNA | AY093206.1 | - | 1 - 2 | 694 | 117 | |
| BT008471.1 | - | 1 - 2 | 348 | 117 | ||
More Information
Set Membership: |
|

AT-HSFB2B
Genomic Map