Assay Details
| Gene Symbol: HAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117241.2 | NP_192909.2 | 14 - 15 | 1535 | 94 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  Detect Genomic DNA  Arabidopsis thaliana  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 HAP2
HAP2 Genomic Map
Genomic Map Back To Top
 Back To Top