Assay Details
| Gene Symbol: HMGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340735.1 | NP_001319906.1 | 7 - 8 | 1081 | 116 | |
| NM_117251.3 | NP_192919.1 | 7 - 8 | 961 | 116 | ||
| NM_179030.1 | NP_849361.1 | 7 - 8 | 734 | 116 | ||
| GenBank mRNA | AY140008.1 | - | 7 - 8 | 734 | 116 | |
| BT008492.1 | - | 5 - 6 | 594 | 116 | ||
| U79160.1 | - | 7 - 8 | 875 | 116 | ||
More Information
Set Membership: |
|

HMGS
Genomic Map