Assay Details
| Gene Symbol: EMB3006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118055.3 | NP_567582.1 | 7 - 8 | 573 | 73 | |
| GenBank mRNA | AK117802.1 | - | 7 - 8 | 573 | 73 | |
| AY087845.1 | - | 7 - 8 | 73 | |||
| BT003710.1 | - | 7 - 8 | 492 | 73 | ||
More Information
Set Membership: |
|

EMB3006
Genomic Map