Assay Details
Gene Symbol: BGAL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343683.1 | NP_001330417.1 | 2 - 3 | 459 | 93 | |
NM_122078.5 | NP_568399.4 | 5 - 6 | 1053 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
