Assay Details
Gene Symbol: BGAL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343683.1 | NP_001330417.1 | 3 - 4 | 578 | 90 | |
NM_122078.5 | NP_568399.4 | 6 - 7 | 1172 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
