Assay Details
Gene Symbol: SUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036838.2 | NP_001031915.1 | 8 - 9 | 1467 | 77 | |
NM_001343687.1 | NP_001332603.1 | 8 - 9 | 1267 | 77 | ||
NM_122090.4 | NP_197583.1 | 7 - 8 | 1866 | 77 | ||
GenBank mRNA | AK316826.1 | - | 7 - 8 | 1273 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
