Assay Details
Gene Symbol: AIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122099.4 | NP_197591.1 | 2 - 3 | 559 | 84 | |
GenBank mRNA | AY081329.1 | - | 2 - 3 | 498 | 84 | |
BT000238.1 | - | 2 - 3 | 366 | 84 | ||
DQ059130.1 | - | 2 - 3 | 366 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
