Assay Details
| Gene Symbol: B73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343692.1 | NP_001332397.1 | 9 - 10 | 2231 | 76 | |
| NM_001343693.1 | NP_001332398.1 | 9 - 10 | 1474 | 76 | ||
| NM_122108.4 | NP_197599.1 | 10 - 11 | 1734 | 76 | ||
More Information
Set Membership: |
|

B73
Genomic Map