Assay Details
| Gene Symbol: B73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343692.1 | NP_001332397.1 | 1 - 2 | 309 | 75 | |
| NM_122108.4 | NP_197599.1 | 1 - 2 | 319 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001343692.1 transcripts.
Set Membership: |
|

B73
Genomic Map