Assay Details
| Gene Symbol: AGO9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122122.3 | NP_197613.2 | 22 - 23 | 2633 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGO9| Gene Symbol: AGO9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122122.3 | NP_197613.2 | 22 - 23 | 2633 | 82 | |
Set Membership: |
|