Assay Details
Gene Symbol: NCLPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122220.5 | NP_568427.1 | 1 - 2 | 448 | 89 | |
GenBank mRNA | AK118523.1 | - | 1 - 2 | 363 | 89 | |
BT005261.1 | - | 1 - 2 | 310 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
