Assay Details
| Gene Symbol: PYRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122236.3 | NP_568428.1 | 6 - 7 | 1189 | 64 | |
| GenBank mRNA | AK228165.1 | - | 6 - 7 | 1189 | 64 | |
| BT026524.1 | - | 6 - 7 | 1028 | 64 | ||
More Information
Set Membership: |
|

PYRD
Genomic Map