Assay Details
Gene Symbol: STE14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343798.1 | NP_001330699.1 | 3 | 513 | 61 | |
NM_122238.2 | NP_197723.1 | 1 | 262 | 61 | ||
GenBank mRNA | DQ446977.1 | - | 1 | 262 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
