Assay Details
| Gene Symbol: AT5G23710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122276.3 | NP_197760.1 | 2 | 706 | 71 | |
| GenBank mRNA | BT010606.1 | - | 1 | 567 | 71 | |
| Gene Symbol: AT5G23700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343819.1 | NP_001330555.1 | 17 | 1962 | 71 | |
More Information
Set Membership: |
|

AT5G23710
Genomic Map