Assay Details
Gene Symbol: AT5G23850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122290.4 | NP_197774.1 | 5 - 6 | 1596 | 80 | |
GenBank mRNA | AK221455.1 | - | 5 - 6 | 1342 | 80 | |
BT014854.1 | - | 5 - 6 | 1268 | 80 | ||
KJ138669.1 | - | 5 - 6 | 1268 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
