Assay Details
Gene Symbol: SS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203449.1 | NP_001190378.1 | 6 - 7 | 1285 | 74 | |
NM_122336.5 | NP_197818.1 | 6 - 7 | 1509 | 74 | ||
GenBank mRNA | AK226881.1 | - | 6 - 7 | 1218 | 74 | |
AY128273.1 | - | 6 - 7 | 1207 | 74 | ||
KJ138668.1 | - | 6 - 7 | 1141 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
