Assay Details
| Gene Symbol: IRE1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203453.2 | NP_001190382.1 | 2 - 3 | 677 | 74 | |
| NM_001203454.1 | NP_001190383.1 | 3 - 4 | 614 | 74 | ||
| NM_122344.5 | NP_568444.1 | 2 - 3 | 620 | 74 | ||
| GenBank mRNA | AY057480.1 | - | 2 - 3 | 565 | 74 | |
| AY057897.1 | - | 2 - 3 | 530 | 74 | ||
More Information
Set Membership: |
|

IRE1-1
Genomic Map