Assay Details
| Gene Symbol: BIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161263.1 | NP_001154735.1 | 5 - 6 | 877 | 80 | |
| NM_001161264.1 | NP_001154736.1 | 5 - 6 | 956 | 80 | ||
| NM_001161265.1 | NP_001154737.1 | 5 - 6 | 944 | 80 | ||
| NM_001161266.1 | NP_001154738.1 | 5 - 6 | 944 | 80 | ||
| NM_001161267.1 | NP_001154739.1 | 5 - 6 | 968 | 80 | ||
| NM_001343888.1 | NP_001330967.1 | 4 - 5 | 1073 | 80 | ||
| NM_001343890.1 | NP_001330965.1 | 5 - 6 | 944 | 80 | ||
| NM_122371.5 | NP_197851.5 | 5 - 6 | 947 | 80 | ||
More Information
Set Membership: |
|

BIN4
Genomic Map