Assay Details
Gene Symbol: BIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161263.1 | NP_001154735.1 | 8 - 9 | 1089 | 101 | |
NM_001161264.1 | NP_001154736.1 | 8 - 9 | 1222 | 101 | ||
NM_001161265.1 | NP_001154737.1 | 8 - 9 | 1210 | 101 | ||
NM_001161266.1 | NP_001154738.1 | 8 - 9 | 1198 | 101 | ||
NM_001161267.1 | NP_001154739.1 | 8 - 9 | 1234 | 101 | ||
NM_001343885.1 | NP_001330963.1 | 8 - 9 | 1192 | 101 | ||
NM_001343886.1 | NP_001330964.1 | 7 - 8 | 1107 | 101 | ||
NM_001343887.1 | NP_001330968.1 | 7 - 8 | 1096 | 101 | ||
NM_001343888.1 | NP_001330967.1 | 7 - 8 | 1339 | 101 | ||
NM_001343889.1 | NP_001330966.1 | 6 - 7 | 1213 | 101 | ||
NM_001343890.1 | NP_001330965.1 | 8 - 9 | 1204 | 101 | ||
NM_122371.5 | NP_197851.5 | 8 - 9 | 1213 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
