Assay Details
| Gene Symbol: CRY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122394.4 | NP_568461.3 | 8 - 9 | 1236 | 159 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRY3| Gene Symbol: CRY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122394.4 | NP_568461.3 | 8 - 9 | 1236 | 159 | |
Set Membership: | 
		   
					
						
							 |