Assay Details
Gene Symbol: AT5G26260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122527.4 | NP_568483.1 | 2 - 3 | 368 | 80 | |
GenBank mRNA | AF370133.1 | - | 2 - 3 | 160 | 80 | |
AY051023.1 | - | 2 - 3 | 147 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
