Assay Details
| Gene Symbol: AGL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343985.1 | NP_001318659.1 | 1 - 2 | 294 | 80 | |
| GenBank mRNA | AK226438.1 | - | 1 - 2 | 274 | 80 | |
| AK227520.1 | - | 1 - 2 | 282 | 80 | ||
| BT010906.1 | - | 1 - 2 | 263 | 80 | ||
More Information
Set Membership: |
|

AGL26
Genomic Map