Assay Details
| Gene Symbol: AGL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343985.1 | NP_001318659.1 | 5 - 6 | 542 | 93 | |
| GenBank mRNA | AK226438.1 | - | 5 - 6 | 522 | 93 | |
| AK227520.1 | - | 5 - 6 | 530 | 93 | ||
| BT010906.1 | - | 5 - 6 | 511 | 93 | ||
More Information
Set Membership: |
|

AGL26
Genomic Map