Assay Details
Gene Symbol: SYP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036873.2 | NP_001031950.1 | 3 - 4 | 1058 | 135 | |
NM_001343989.1 | NP_001331924.1 | 3 - 4 | 797 | 135 | ||
NM_122580.4 | NP_198050.1 | 3 - 4 | 1058 | 135 | ||
GenBank mRNA | AF067789.2 | - | 3 - 4 | 651 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
