Assay Details
| Gene Symbol: TPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122585.5 | NP_198055.3 | 10 - 11 | 1564 | 86 | |
| GenBank mRNA | BT004037.1 | - | 10 - 11 | 1395 | 86 | |
| BT005165.1 | - | 10 - 11 | 1262 | 86 | ||
More Information
Set Membership: |
|

TPR3
Genomic Map