Assay Details
Gene Symbol: TPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203480.1 | NP_001190409.1 | 1 - 2 | 222 | 145 | |
NM_122585.5 | NP_198055.3 | 1 - 2 | 380 | 145 | ||
GenBank mRNA | BT004037.1 | - | 1 - 2 | 211 | 145 | |
BT005165.1 | - | 1 - 2 | 78 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
