Assay Details
| Gene Symbol: AGL104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102063.3 | NP_173632.1 | 7 - 8 | 534 | 73 | |
| GenBank mRNA | DQ056457.1 | - | 7 - 8 | 448 | 73 | |
More Information
Set Membership: |
|

AGL104
Genomic Map