Assay Details
| Gene Symbol: UGT85A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036001.1 | NP_001031078.1 | 1 - 2 | 567 | 126 | |
| NM_102086.3 | NP_173653.1 | 1 - 2 | 737 | 126 | ||
| GenBank mRNA | AB016819.1 | - | 1 - 2 | 534 | 126 | |
| AF332418.1 | - | 1 - 2 | 503 | 126 | ||
More Information
Set Membership: |
|

UGT85A2
Genomic Map