Assay Details
| Gene Symbol: UGT85A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102089.5 | NP_173656.1 | 1 - 2 | 876 | 99 | |
| GenBank mRNA | AY081339.1 | - | 1 - 2 | 580 | 99 | |
| BT008765.1 | - | 1 - 2 | 514 | 99 | ||
| KJ138990.1 | - | 1 - 2 | 514 | 99 | ||
More Information
Set Membership: |
|

UGT85A1
Genomic Map