Assay Details
Gene Symbol: PAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102123.4 | NP_173690.1 | 2 - 3 | 1363 | 136 | |
GenBank mRNA | AY054490.1 | - | 2 - 3 | 1362 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
