Assay Details
| Gene Symbol: AT1G22950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102142.5 | NP_173708.3 | 2 - 3 | 476 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G22950| Gene Symbol: AT1G22950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102142.5 | NP_173708.3 | 2 - 3 | 476 | 102 | |
Set Membership: |
|