Assay Details
Gene Symbol: AST91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332587.1 | NP_001320231.1 | 1 - 2 | 694 | 73 | |
NM_102157.4 | NP_173722.1 | 2 - 3 | 578 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001332587.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
