Assay Details
Gene Symbol: AST91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332586.1 | NP_001320230.1 | 4 - 5 | 1335 | 105 | |
NM_001332587.1 | NP_001320231.1 | 5 - 6 | 1250 | 105 | ||
NM_001332588.1 | NP_001320232.1 | 5 - 6 | 1205 | 105 | ||
NM_102157.4 | NP_173722.1 | 6 - 7 | 1134 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
