Assay Details
Gene Symbol: AST91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332586.1 | NP_001320230.1 | 6 - 7 | 2065 | 83 | |
NM_001332587.1 | NP_001320231.1 | 7 - 8 | 1980 | 83 | ||
NM_001332588.1 | NP_001320232.1 | 7 - 8 | 1935 | 83 | ||
NM_102157.4 | NP_173722.1 | 8 - 9 | 1864 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
