Assay Details
| Gene Symbol: RPL27AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102178.3 | NP_173743.1 | 1 | 595 | 148 | |
| Gene Symbol: AT1G23300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102179.2 | NP_173744.1 | 9 | 1856 | 148 | |
More Information
Set Membership: |
|

RPL27AB
Genomic Map