Assay Details
| Gene Symbol: AT1G24210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102267.3 | NP_564212.1 | 2 - 3 | 273 | 131 | |
| GenBank mRNA | AY086413.1 | - | 2 - 3 | 245 | 131 | |
| BT024693.1 | - | 1 - 2 | 133 | 131 | ||
More Information
Set Membership: |
|

AT1G24210
Genomic Map