Assay Details
| Gene Symbol: RBL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198164.2 | NP_001185093.1 | 3 - 4 | 599 | 67 | |
| NM_102339.3 | NP_173900.2 | 3 - 4 | 515 | 67 | ||
More Information
Set Membership: |
|

RBL10
Genomic Map