Assay Details
Gene Symbol: PFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084127.1 | NP_001077596.1 | 9 - 10 | 1918 | 63 | |
NM_102365.5 | NP_173925.3 | 10 - 11 | 2104 | 63 | ||
GenBank mRNA | AY170377.1 | - | 10 - 11 | 1735 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
