Assay Details
Gene Symbol: PFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084127.1 | NP_001077596.1 | 11 - 12 | 2021 | 70 | |
NM_102365.5 | NP_173925.3 | 12 - 13 | 2207 | 70 | ||
GenBank mRNA | AY170377.1 | - | 12 - 13 | 1838 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
