Assay Details
Gene Symbol: PFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102365.5 | NP_173925.3 | 1 - 2 | 469 | 76 | |
GenBank mRNA | AY170377.1 | - | 1 - 2 | 100 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
