Assay Details
| Gene Symbol: AT1G26190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332692.1 | NP_001323084.1 | 9 - 10 | 1624 | 97 | |
| NM_001332693.1 | NP_001323086.1 | 9 - 10 | 1624 | 97 | ||
| NM_001332694.1 | NP_001323087.1 | 10 - 11 | 1521 | 97 | ||
| NM_001332695.1 | NP_001323085.1 | 9 - 10 | 1606 | 97 | ||
| NM_102384.4 | NP_173944.1 | 9 - 10 | 1660 | 97 | ||
| GenBank mRNA | AY091017.1 | - | 9 - 10 | 1606 | 97 | |
| AY117297.1 | - | 8 - 9 | 1385 | 97 | ||
More Information
Set Membership: |
|

AT1G26190
Genomic Map