Assay Details
| Gene Symbol: BGLU40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102418.4 | NP_173978.1 | 3 - 4 | 654 | 89 | |
| GenBank mRNA | AY045927.1 | - | 3 - 4 | 488 | 89 | |
More Information
Set Membership: |
|

BGLU40
Genomic Map