Assay Details
| Gene Symbol: BGLU40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102418.4 | NP_173978.1 | 7 - 8 | 1186 | 83 | |
| GenBank mRNA | AY045927.1 | - | 7 - 8 | 1020 | 83 | |
| AY142610.1 | - | 7 - 8 | 885 | 83 | ||
More Information
Set Membership: |
|

BGLU40
Genomic Map